ContactUsLinkComponent
VWR® Laboratory LED Zoom Stereo Series
VWR® Laboratory LED Zoom Stereo Series
Supplier:  VWR International
Certificates
VWR® Laboratory LED Zoom Stereo Series
VWR® Laboratory LED Zoom Stereo Series
Supplier:  VWR International
Certificates
Specifications
  • Catalog No:
  • 10836-006
  • 10836-004
  • Head:
  • 35° inclined
  • 35° inclined
  • Eyepieces:
  • WF 10×/23 mm
  • WF 10×/23 mm
  • Magnification:
  • 0.75× – 5×
  • 0.75× – 5×
  • Zoom lens:
  • 6,7:1
  • 6,7:1
  • Working distance:
  • 113 mm
  • 113 mm
  • Illumination:
  • 3W Intensity controlled transmitted LED illumination plus 3W Incident LED illumination assembly
  • 3W Intensity controlled transmitted LED illumination plus 3W Incident LED illumination assembly
  • Stand:
  • Transmitted large working area stand
  • Transmitted large working area stand
  • Electrical:
  • 100 – 240V
  • 100 – 240V
  • Optical design:
  • Greenough Zoom
  • Greenough Zoom