Avantor® Genesis, C4-300 HPLC Columns
# de catalogue 75813-106
Fournisseur: HICHROM LIMITED
Spécifications
- Column Format:Semi-preparative
- I.D.:10.00 mm
- Length:250 mm
- Particle size:4 µm
- Column Hardware:Stainless Steel
- Cat. no.:75813-106
- Packing Material:Silica
- Phase:C4-300
- Supplier No.:8P25976
Spécifications
A propos de cet article
Avantor® Genesis phases are based on high purity metal-free spherical silica.
- Good peak shape and reproducibility
- Long column lifetime
- pH stability 1 to 10
They are suitable for the analysis of a wide range of analytes.
Customers Who Bought This Also Bought