Spécifications
- Description:Disposable Multi-Well Embedding Mold
- Cat. no.:103301-800
- Supplier No.:70918
Spécifications
A propos de cet article
Disposable flexible LDPE mold for embedding.
A clear flexible LDPE mold to clean the molds. 2 troughs: 1 L x 1/8 W x 1/8" D. 14 cavities: 1/4 L x 1/2 W x 1/8" D. 1 trough: 23/4 L x 3/32 W x 1/8 " D. 10 cavities: 3/8 L x 3/4 W x 1/8" D.