To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IL-12/IL-23 p40
- Antigen symbol:IL-12/IL-23 p40 (C17.8)
- Clonality:Monoclonal
- Clone:C17.8
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:25 ug
- Antigen synonyms:NKSF|Interleukin-23|IL12|CLMF|Cytotoxic lymphocyte maturation factor|CTL maturation factor|T-cell stimulating factor|TcMF|TSF|Natural killer cell stimulatory factor|IL23 p40|Interleukin-12
- Form:liquid
- Cat. no.:75842-596
- Supplier No.:83312-60-25
Specifications
About this item
The C17.8 antibody specifically reacts with the p40 subunit of IL-12, as free monomer, as a homodimer, or as a part of the p70 dimer. C17.8 seems to neutralize the activity of mouse IL-12 and IL-23. IL-12 is the p70 heterodimer of p35 and p40, expressed by the macrophages, dendritic cells, and monocytes. The C17.8 antibody cross-reacts with IL-23, since IL-23 also contains one p40 subunit.
Type: Primary
Antigen: IL-12/IL-23 p40 (C17.8)
Clonality: Monoclonal
Clone: C17.8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse