To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Clonality:Monoclonal
- Clone:EOS9.1
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Antigen symbol:CD95
- Flow cytometry:Yes
- Size:50 μg
- Format:SAFIRE Purified
- Form:liquid
- Antigen synonyms:Fas/APO-1|APO-1|TNFRSF6|Fas
- Cat. no.:75844-192
- Supplier No.:08011-25-50
Spécifications
A propos de cet article
The EOS9.1 monoclonal antibody specifically reacts with human CD95, a 45 kDA type I membrane molecule. CD95 is a member of the tumor necrosis factor receptor (TNF-R) family and is involved in the mechanisms that lead to cellular apoptosis. It is expressed on fibroblasts, monocytes, lymphocytes, and neutrophils. CD95 induces apoptosis after binding with CD178 or Fas ligand and the EOS9.1 antibody is reported to be used for inducing apoptosis in Fas-positive cells.
Type: Primary
Antigen: CD95
Clonality: Monoclonal
Clone: EOS9.1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM, kappa
Reactivity: Human