Anti-CD3 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: Hit3a]
Catalogue # 75841-660
Fournisseur: Biogems
Spécifications
- Antibody type:Primary
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:Hit3a
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:100 tests
- Form:liquid
- Antigen synonyms:Leu-4|T3
- Cat. no.:75841-660
- Supplier No.:05111-60-100
Spécifications
À propos de cet article
The Hit3a monoclonal antibody specifically reacts with the ε chain of the CD3/T lymphocyte antigen receptor complex. The CD3 complex is part of the TCR complex, expressed by all mature T lymphocytes and by the thymocyte lineage. CD3 enhances the antigen recognition by signal transduction.The HIT3a antibody cross-links with the TCR complex, initiating the cellular activation and proliferation, but it cannot be used for intracellular CD3 staining, as it is able to stain only the surface CD3 complex.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: Hit3a
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human