To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen symbol:CD197
- Clonality:Monoclonal
- Clone:3D12
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100ug
- Antigen synonyms:EBI-1|MIP-3 beta Receptor
- Form:liquid
- Cat. no.:75843-166
- Supplier No.:20011-20-100
Specifications
About this item
The 3D12 monoclonal antibody specifically reacts with human CD197 (CCR7 or EBI-1), a seven-transmembrane G-protein-coupled. It is the receptor for the secondary lymphoid-tissue chemokine (SLC or 6CKine), CCL19, and CCL21. CD197 is expressed on subsets of B and T cells, and mature dendritic cells. The monoclonal 3D12 antibody recognizes the N-terminus of the receptor.
We recommend that this antibody be incubated for at least 45 minutes at 4°C for optimal staining.
Type: Primary
Antigen: CD197
Clonality: Monoclonal
Clone: 3D12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human