To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Fas Ligand
- Antigen symbol:CD178
- Clonality:Monoclonal
- Clone:NOK-1
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:25 μg
- Antigen synonyms:FasL|CD95L|Fas Ligand
- Form:liquid
- Cat. no.:75843-188
- Supplier No.:20811-30-25
Specifications
About this item
The NOK-1 monoclonal antibody specifically reacts with human CD178, which is the CD95 or Fas ligand. CD178 is a TNF superfamily type II transmembrane glycoprotein expressed by activated T and NK cells and is involved in Fas-mediated apoptosis of lymphocytes. CD178 is also expressed by monocytes, neutrophils, granulocytes and the parenchymal cells of the retina and cornea. The NOK-1 antibody has been reported to bind to COOH-terminus of the Fas ligand in the region associated with Fas binding.
Type: Primary
Antigen: CD178
Clonality: Monoclonal
Clone: NOK-1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human