Hydrion™ pH Test Paper Refills for Dispensers, Micro Essential Laboratory
Fournisseur: Micro Essential
Certificats
A propos de cet article
Hydrion™ pH test paper refills are used for Hydrion™ dispensers.
- Ideal for double-roll and single-roll dispensers
- Standard-size rolls: 4.6 m x 0.53 cm
Come in pack of five rolls; each pack is a tightly sealed plastic container, complete with two color charts.
Delivery: Includes colorimetric chart.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.