To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Protein Tyrosine Phosphatase Receptor Type C
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:CD45
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:100 µg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
- Amino acid number:1113 to 1304
- Storage temperature:–20 °C
- Immunogen:E.coli-derived human CD45 recombinant protein (Position: R1113-S1304). Human CD45 shares 68% amino acid (aa) sequences identity with both mouse and rat CD45.
- Purification:purified by immunogen affinity purification
- Cat. no.:10207-470
- Supplier No.:PB9096
Spécifications
A propos de cet article
Polyclonal antibody for CD45/PTPRC detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. CD45/PTPRC information: Molecular Weight: 147254 MW; Subcellular Localization: Membrane ; Single-pass type I membrane protein . Membrane raft . Colocalized with DPP4 in membrane rafts.
Type: Primary
Antigen: CD45
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human