To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:CMA1 Picoband™
- Antigen symbol:CMA1/Chymase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Immunogen:E.coli-derived human CMA1 recombinant protein (Position: I22-N247). Human CMA1 shares 75% and 74% amino acid (aa) sequence identity with mouse and rat CMA1, respectively.
- Amino acid number:22 to 247
- Purification:purified by immunogen affinity purification
- Storage temperature:–20 °C
- Form:Lyophilized
- Cross adsorption:No
- Cat. no.:10207-444
- Supplier No.:PB9055
Specifications
About this item
Polyclonal antibody for Chymase/CMA1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. Chymase/CMA1 information: Molecular Weight: 27325 MW; Subcellular Localization: Secreted. Cytoplasmic granule. Mast cell granules; Tissue Specificity: Mast cells in lung, heart, skin and placenta. Expressed in both normal skin and in urticaria pigmentosa lesions.
Type: Primary
Antigen: CMA1/Chymase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human