Avantor® Alltima, Phenyl HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L11
Less hydrophobic than C18 phase. Selective for aromatic compounds.
Spécifications
- Catalog No:
- 75797-302
- 75797-300
- 75797-304
- 75797-286
- 75797-284
- 75797-290
- 75797-288
- 75797-294
- 75797-298
- 76398-446
- 75797-282
- 75830-850
- 75830-372
- Description:
- 75797-304
- All-Guard cartridge for 3.0 – 4.6 mm ID columns (Use with holder 80101/N and coupler HI-081)
- All-Guard cartridge for 2.1 mm ID columns (Use with holder 80101/N and coupler HI-081)
- Carbon load:
- 7.5%
- Endcapped:
- Yes
- Surface area:
- 340 m2/g
- USP code:
- L11