Avantor® Alltima, Cyano HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L10
General purpose cyano suitable for normal or reversed phase use. Suitable for rugged normal phase applications.
Spécifications
- Catalog No:
- 75797-268
- 75797-274
- 75797-272
- 75797-278
- 75797-276
- 75797-280
- 75797-264
- 75830-854
- 75797-266
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- 340 m²/g
- USP code:
- L10
- L10
- L10
- L10
- L10
- L10
- L10
- L10