Avantor® Alltima C18-LL, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L1
Lower carbon load than traditional Alltima C18. Reversed phase applications that require a less hydrophobic C18 phase.
Customers Who Bought This Also Bought