Avantor® Alltima, Amino HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Alltima HPLC columns are high quality phases designed for general purpose applications.
- Base deactivated silica phases
- Stable bonding for long column lifetime
- Multiple selectivity options
- USP L8
General purpose amino suitable for normal or reversed phase use. Recommended for carbohydrate analysis or as a weak anion exchanger.
Customers Who Bought This Also Bought