Avantor® Genesis, Silica HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Genesis phases are based on high purity metal-free spherical silica.
- Good peak shape and reproducibility
- Long column lifetime
- pH stability 1 to 10
They are suitable for the analysis of a wide range of analytes.
Customers Who Bought This Also Bought