SW® PowerForm® PF-95BK Black 5.6 mil Sustainable Nitrile Exam Gloves
Fournisseur: SW Safety Solutions
Certificats
A propos de cet article
These gloves protect hands and the environment, with a sustainable single-use design that combines performance, durability, and tactile sensitivity for critical tasks.
- EcoTek® sustainable technology
- Consistent thickness for greater dexterity and tactile sensitivity
- Fully textured for excellent wet and dry grip
- Silicone-free for critical environments
- Clean processing technology reduces potential skin irritation
- Optimal thickness for durability and tactile sensitivity
- Tear-resistant beaded cuff eases donning
- 1.5 AQL exceeds laboratory standards
EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Certifications: WBENC Women Owned; NCMSDC Minority Business; Small Business Enterprise; CE 0493. PowerForm® is dermatalogically approved by the Skin Health Alliance (SHA), and meets stringent standards that ensure skin safety.
Ordering information: Glove sizes are as follows; S (6.5 to 7), M (7.5 to 8), L (8.5 to 9), XL (9.5 to 10), XXL (10.5 to 11).
Spécifications
- Catalog No:
- 76051-106
- 76051-114
- 76051-112
- 76051-110
- 76051-108
- Model:
- PF-95BK
- PF-95BK
- PF-95BK
- PF-95BK
- PF-95BK
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes