A propos de cet article
These gloves combine sweat management and unprecedented strength in a fully-textured glove that delivers the ultimate in comfort and performance.
- DriTek® comfort technology with absorbent lining that wicks away moisture for added extended use
- EcoTek® sustainable technology
- ANSI/EN level 3 abrasion resistance for durability and extended wear
- Fully textured surface for excellent grip
- Optimal thickness for durability and tactile sensitivity
- 1.5 AQL exceeds laboratory standards
- Up to 3× longer lasting than standard nitrile gloves
- SHA dermatologically approved
Now with EcoTek® sustainable technology. Additional features include an extended-cuff for splash protection and ANSI/EN level 3 abrasion resistance for a nitrile glove that lasts three times longer than standard gloves. ASTM tested against viral penetration and low dermatitis potential. EcoTek® gloves tested to ASTM D5526-12 standard test method for determining anaerobic biodegradation of plastic materials under accelerated landfill conditions. Results are 92.6% biodegradation in 60% solids landfill at 945 days. Future results cannot be predicted/extrapolated. For additional information on the test reports, contact [email protected].
Spécifications
- Catalog No:
- 76051-216
- 76051-214
- 76051-212
- 76051-154
- 76051-152
- 76051-218
- 76051-210
- Model:
- MM-11BK
- MM-11BK
- MM-11BK
- MM-11BK
- MM-11BK
- MM-11BK
- MM-11BK
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- EN norm:
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- EN ISO 21420:2020; EN 455-2000-2015
- Food contact approved:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes