Septa for 10-425 Screw-Thread Caps, MicroLiter
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Septa for the original wide-opening vials from MicroLiter are ideal for chromatography applications where vials remain stationary.
Packaging: Class 10000 Clean Room Packaging includes Dispenser Pack septa for proper storage and handling on the bench.
Customers Who Bought This Also Bought