WHEATON® MicroLiter Septa for Screw-Thread Caps, DWK Life Sciences
Supplier: Avantor
Certificates
About this item
Septa for the original screw-thread autosampler caps and vials from MicroLiter offer convenient packaging, which includes all of the enhanced components and configurations needed to fit your workflow.
Packaging: Class 10000 Clean Room Packaging includes Dispenser Pack septa for proper storage and handling on the bench.
Customers Who Bought This Also Bought