Septa for 9 mm Screw-Thread Caps, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
MicroLiter's 9 mm screw-thread caps are available in multiple colors and septa types.
- ScewTin™ Ring is a metallic disc for x-y-z robotics that employ magnetic capture to move vials
- Post Baked to remove additional siloxanes
Customers Who Bought This Also Bought