Glass Solvent Bottle with ITSP Connector, 1 L, MicroLiter, DWK Life Sciences
# de catalogue 97050-590
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Glass Solvent Bottle
- Cat. no.:97050-590
- Supplier No.:LPAL.1000ML
Spécifications
A propos de cet article
Glass solvent bottle is 1 L and has an 8 mm connector for HTS, HTS-xt, HTC, HTC-xt, HTX, and HTX-xt PAL systems.
Customers Who Bought This Also Bought