Limited Volume Vial Inserts, MicroLiter
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Inserts are made from Type 1 borosilicate glass or polypropylene and meet strict tolerances.
- Conical and flat bottom
- Meet strict tolerances
Conical inserts, inserts with plastic springs, and flat bottom inserts are available to assist in consistent sampling of low-volume samples.
Customers Who Bought This Also Bought