Vydac 208MS C8, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Vydac MS columns are a later development from Grace for the reversed phase HPLC separation of biomolecules.
- Five reversed phase chemistries
- Excellent peak shape with little or no TFA
- High protein recoveries make scale-up easy
- USP L7
Lower hydrophobicity which is better for larger biomolecules. Ideal for biomolecules with 5 to 10K MW.
A proprietary surface treatment and bonding process give Vydac MS columns unique selectivity. A variety of reversed phases makes this product line suitable for the analysis of small peptides to large intact, undenatured proteins.
Vydac is a registered trademark of W.R.Grace and Co.
Customers Who Bought This Also Bought