Boiling Stones, Plain Micro Granules, Hengar
Fournisseur: Hengar
Certificats
A propos de cet article
High purity, white, amphoteric alundum granules.
- Prevent super-heating
Designed for applications involving the boiling of a liquid.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.