Corning® 1536-Well Cyclic Olefin Copolymer Microplates, Corning
Supplier: Corning
Certificates
About this item
1536-Well Cyclic Olefin Copolymer microplates are perfectly suited for high-throughput screening (HTS)
- Bar coded on the short side of the microplate
- Cyclic Olefin Copolymer (COC) material
- Flat top for improved sealing
- Ultra clear 127 µm film thickness
- Variety of treatment options
- Kalypsys and GNF system compatible
The ultraclear, 127 μm film bottom thickness is ideal for cell-based assays that require advanced optical analysis. The COC microplates are mechanically stable and inherently resistant to a broad range of solvents including DMSO and alcohols, allowing for compound storage as well.
A variety of surface treatments enable both cell-based and biochemical assays.
Specifications
- Catalog No:
- 76175-642
- 76175-640
- 76175-638
- 76175-644
- 76175-636
- 76175-634
- 76175-632
- 76175-630
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk