Unifrost™ Adhesive Microscope Slides, Azer Scientific
# de catalogue 76076-740
Fournisseur: Azer Scientific Inc.
Spécifications
- Color:White
- Description:Adhesive microscope slide
- L×W:75×25" mm (3×1")
- Thickness:1 mm
- Cat. no.:76076-740
Spécifications
A propos de cet article
These adhesive microscope slides are coated with Poly-L-Lysine.
- Alternative to silane coated slides
- Ideal for IHC procedures
- Designed for the toughest, most difficult procedures
Permanent adhesive firmly anchors all specimen types to the slides.
Customers Who Bought This Also Bought