Student Inorganic and Organic Molecular Model Set, 65 Pieces
# de catalogue 470231-426
Fournisseur: Wards
Spécifications
- Description:Student Molecular Model Set, 65 Pieces
- Cat. no.:470231-426
Spécifications
A propos de cet article
Perfect Starter Set
- 32 atoms
- 33 bonds
Contents: 6 Black Carbon (4 Hole, 22 mm), 10 White Hydrogen (1 Hole, 15 mm), 4 Red Oxygen (2 Hole, 20 mm), 2 Blue Nitrogen (4 Hole, 22 mm), 2 Green Halogen (1 Hole, 20 mm), 2 Green Halogen (4 Hole, 22 mm), 1 Yellow Sulfur (2 Hole, 22 mm), 1 Yellow Sulfur (4 hole, 22 mm), 1 Purple Phosphorus (4 Hole, 22 mm), 2 Grey Metal (1 Hole, 15 mm), 1 Grey Metal (2 Hole, 22 mm), 8 Long Grey Bonds, 25 medium Grey Bonds.
Customers Who Bought This Also Bought