Molecular Model Set, 92 Atoms
# de catalogue 470231-436
Fournisseur: Wards
Spécifications
- Description:Molecular Model Set, 92 Atoms
- Cat. no.:470231-436
Spécifications
A propos de cet article
Advanced Model Set
- 92 atoms
- 234 links
Contents: 36 White 1 Face Hydrogen (30 mm), 18 Black 4 Face Carbon (38 mm), 8 Black 3 Face Carbon (38 mm), 2 Black 2 Face Carbon (38 mm), 2 Blue 4 Face Nitrogen (38 mm), 6 Blue 4 Face Nitrogen (38 mm), 1 Blue 1 Face Nitrogen (38 mm), 8 Red 2 Face Oxygen (38 mm), 2 Red 1 Face Oxygen (38 mm), 1 Light Blue 1 Face Fluorine (38 mm), 6 Green 1 Face Chlorine (50 mm) 1 Brown 1 Face Bromine (50 mm), 1 Pink 1 Face Iodine (50 mm), 40 Medium Links (50 mm), 64 Long Links (65 mm), 4 Flexible Links (65 mm), 6 Turning Links (70 mm), 120 Small Links (16 mm).
Customers Who Bought This Also Bought