Molecular Model Set, 104 Atoms
# de catalogue 470231-434
Fournisseur: Wards
Spécifications
- Description:Molecular Model Set, 104 Atoms
- Cat. no.:470231-434
Spécifications
A propos de cet article
Comprehensive Model Set
- 99 atoms
- 238 links
Contents: 40 White 1 face Hydrogen, 20 Black 4 Face Carbon, 10 Black 3 Face Carbon, 4 Black 2 Face Carbon, 6 Red 1 Face Oxygen, 10 Red 2 Face Oxygen , 1 Blue 4 Face Nitrogen, 3 Blue 3 Face Nitrogen, 4 Blue 2 Face Nitrogen, 2 Green 1 Face Chlorine, 2 Light Blue 1 Face Fluorine, 2 Brown 1 Face Bromine, 48 Large Connector for Larger Holes, 48 Medium Connector for Larger Holes, 120 Small Connectors for Larger Holes, 16 Connectors for Smaller holes, 6 Rotating Connectors.
Customers Who Bought This Also Bought