A propos de cet article
Built to handle daily routine use by universities, clinics, and labs, these upright compound microscopes utilize high quality components and a sturdy construction. From the high-end optics included in the eyepieces and objectives to the robust stage and base, these microscopes are built to last through consistent use of demanding medical, and life-science research applications.
- 3W LED illumination
- Vivid and detailed images with optimal image contrast are achieved via included plan achromat objectives with multi-layer lens coatings
- A hard coated and chemical resistant stage ensures long term durability, while its 76 x 50 mm travel range provides great flexibility
- Eyepiece tubes have been optimized for ergonomic perfection
- With a viewing angle of 30° and an interpupillary distance of 55 – 75 mm, users can enjoy hours of fatigue free microscope use
- A full Koehler feature enables the condenser to be set at any height
- Student proofing features include lockable eyepieces to prevent unwanted removal
The VWR® Professional Plus Series is purpose-built to perform consistently through the course of daily routine use
Its durable construction and quality materials prepare it for a long lifetime, taking on the demanding applications of universities, clinics, and labs. From the eyepieces to the objectives, the VWR® Professional Plus Series contains high-end optical components to deliver the supreme optical performance required by professional microscope users. EC Plan Achromat objectives, together with a calculated tube lens, yield fully corrected, perfected intermediate images without colored fringes. These objectives are also multi-layer coated for improved contrast, enhancing even the weakest slide stainings to their maximum level of detail.
The ergonomic construction of the VWR® Professional Plus Series means users can enjoy hours of viewing, free of fatigue and eye-strain. With a 30° viewing angle and an interpupillary distance of 55 – 75 mm, along with a large field of view, slide after slide can be viewed with speed and comfort.
The stage offers a 76 x 50 mm range and a stage area of 175 x 40 mm, giving the user a high degree of freedom of movement, while the slide holder is designed to provide movement consistency. The stage includes a hard coated and chemically resistant surface, keeping it safe from contamination and abrasion during routine use.
With all this and more, the VWR® Professional Plus Series is a durable, high quality upright compound microscope, fully capable of living up to the ongoing demands of professionals with applications in life-science, universities and clinics.
Spécifications
- Catalog No:
- 76122-380
- 76122-382
- 76122-408
- 76122-406
- Model:
- VWR® Professional Plus
- VWR® Professional Plus Phase
- VWR® Professional Plus
- VWR® Professional Plus Phase
- Head:
- Siedentopf, 30° Inclined, 360° Rotating, 20:80 Light Split
- Siedentopf, 30° Inclined, 360° Rotating
- Siedentopf, 30° Inclined, 360° Rotating, 20:80 Light Split
- Siedentopf, 30° Inclined, 360° Rotating
- Eyepieces:
- Widefield high eyepoint eyepiece N-WF10X/20 mm with diopter adjustment on both eyepieces, with rubber eyecup (paired)
- Widefield high eyepoint eyepiece N-WF10X/20 mm with diopter adjustment on both eyepieces, with rubber eyecup (paired)
- Widefield high eyepoint eyepiece N-WF10X/20 mm with diopter adjustment on both eyepieces, with rubber eyecup (paired)
- Widefield high eyepoint eyepiece N-WF10X/20 mm with diopter adjustment on both eyepieces, with rubber eyecup (paired)
- Nosepiece:
- Quintuple, Reversed
- Quintuple, Reversed
- Quintuple, Reversed
- Quintuple, Reversed
- Condenser:
- Focusable Abbe condenser N.A. 0.90 with iris diaphragm and slot
- Phase contrast 5 position (BF, DF, PH1, PH2, PH3) turret condenser
- Focusable Abbe condenser N.A. 0.90 with iris diaphragm and slot
- Phase contrast 5 position (BF, DF, PH1, PH2, PH3) turret condenser
- Illumination:
- 3W LED
- 3W LED
- 3W LED
- 3W LED
- Electrical:
- Universal Power Supply 100 – 240 V
- Universal Power Supply 100 – 240 V
- Universal Power Supply 100 – 240 V
- Universal Power Supply 100 – 240 V