Polytron® 12 mm Generator, PT-DA 12/2EC-E157, Kinematica
# de catalogue 97006-686
Fournisseur: Kinematica
Small Business Enterprise
Spécifications
- Description:PT-DA 12 / 2WEC-E157, "W" Style
- Cat. no.:97006-686
- Diameter ×L:12×157 mm
- For Use With:PT 1200 E, PT 1300 D, PT 2500 E
- Sample Volume:2 - 250 ml
- Supplier No.:11030060
- Diameter:12 mm
Spécifications
A propos de cet article
Fits Polyron models, PT 1200 E, PT 1300 D, PT 2500 E as well as discontinued model PT 1600 E.
- Saw tooth stator design for added size reduction effects
- Easy clean design for quick disasembly
- 316L stainless
Precison machined form the finest 316 SS by our skilled Swiss craftsman for years of trouble free service.
Certifications: CE marked.
BodyTextAccessories: Generators and stand with vessel holder sold separately.
Customers Who Bought This Also Bought