ContactUsLinkComponent
Edvotek® InstaStain® Ethidium Bromide Impregnated Cards
Edvotek® InstaStain® Ethidium Bromide Impregnated Cards
# de catalogue 470121-824
Fournisseur:  Edvotek
Numéro CAS:  
INSTASTAIN ETHIDIUM BR 100
Edvotek® InstaStain® Ethidium Bromide Impregnated Cards
# de catalogue 470121-824
Fournisseur:  Edvotek
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    InstaStain® Cards
  • Cat. no.:
    470121-824
  • Supplier No.:
    2002
Spécifications
A propos de cet article
This set contains enough cards to stain 100 gels measuring 7 x 7 cm.
  • Rapid and sensitive
  • Requires UV transilluminator to visualize
  • Less student exposure to Ethidium Bromide than traditional methods
InstaStain® Ethidium Bromide, which contains only a few micrograms of ethidium bromide, is rapid and sensitive. In two minutes, an agarose gel is ready for visualization. Disposal of InstaStain® Ethidium Bromide is minimal compared to the volume of liquid waste generated from the standard ethidium bromide staining procedure.
Customers Who Bought This Also Bought