Spécifications
- Description:InstaStain® Cards
- Cat. no.:470121-824
- Supplier No.:2002
Spécifications
A propos de cet article
This set contains enough cards to stain 100 gels measuring 7 x 7 cm.
- Rapid and sensitive
- Requires UV transilluminator to visualize
- Less student exposure to Ethidium Bromide than traditional methods
InstaStain® Ethidium Bromide, which contains only a few micrograms of ethidium bromide, is rapid and sensitive. In two minutes, an agarose gel is ready for visualization. Disposal of InstaStain® Ethidium Bromide is minimal compared to the volume of liquid waste generated from the standard ethidium bromide staining procedure.