EDVOTEK PCR EdvoBeads™
# de catalogue 470111-884
Fournisseur: Edvotek
Numéro CAS:
Spécifications
- Description:25 EdvoBeads™ Beads
- Cat. no.:470111-884
- Supplier No.:625
Spécifications
A propos de cet article
Simply add your primers and template DNA to a single PCR EdvoBead™ and mix!
- Each vial contains enough beads for 25 PCR reactions
- Stores in ambient temperature
PCR EdvoBead™ provide the reagents for 25 PCR reactions in a convenient ambient-temperature-stable bead. PCR Beads have been optimized for PCR reactions and contain buffer, nucleotides and Taq DNA Polymerase. The only reagents that must be added to the reaction are template DNA and specific primers.
Caution: Each vial contains enough beads for 25 PCR reactions
Stores in ambient temperature
Customers Who Bought This Also Bought