KF Flange Flat Stainless Steel Head, Ace Glass
# de catalogue 76106-990
Fournisseur: Ace Glass
Spécifications
- Description:Flat Stainless Steel Head for use with 10 l Reactors, (1)45/50, (3)29/42, (3)24/40
- Cat. no.:76106-990
- Flange O.D.:200 mm
Spécifications
A propos de cet article
KF Flange flat stainless steel head with holes to accept PTFE standard taper joint inserts.
Customers Who Bought This Also Bought