Deep Vacuum Sublimation Apparatus, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
Improved sublimer designed for deep vacuum, with the vacuum valve on the top of the sublimer to keep the sublimate around the cold finger out of your vacuum trap
- Features a Viton O-Ring and a quick release clamp for deep vacuum operation
- Duran Flange is 150 mm
- Complete item includes outer body, inner cold finger lid, and quick release clamp
- Outer body and Inner cold finger lid also sold as separate items
Increased spacing between the cold finger and outer body leaves more room for your product.
Customers Who Bought This Also Bought