PTFE Bottom Surface Cover for Stainless Steel Flat Head, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
PTFE bottom surface cover will protect stainless steel flat heads from chemical attack.
- Cover made of chemical-resistant PTFE
- Secured by the coupling and the joint inserts
- For use with KF flat flanged stainless steel heads
Customers Who Bought This Also Bought