Rapid-Core Cutting Mat, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
A propos de cet article
Composite cutting mats for coring samples.
- Protects tips and working surface
- Reversible, inert and self-healing
- Sterilized with ethanol
Five layer composite cutting mat will cut cleanly through an object and into the mat. Ensures the cored sample comes out completely without burrs and bits.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.