VWR® Dot Matrix Printer
# de catalogue 10799-132
Fournisseur: VWR International
Spécifications
- Description:VWR® Dot Matrix Printer
- Cat. no.:10799-132
Spécifications
A propos de cet article
Easily obtain results from your VWR® Series Balance with connection to the VWR® Dot Matrix Printer.
- Interface cable for connecting the balance to the printer included with system
- Black print color
- Includes sample roll of printer paper
The printer allows for easy paper replacement with drop in paper loading. With the ability to use 58 mm, 70 mm, or 76 mm paper.
Customers Who Bought This Also Bought