Cubitainer Containers, Low-Density Polyethylene, Paramount Can
Fournisseur: Paramount Can
Certificats
A propos de cet article
Cube shaped inserts designed for use with a corrugated fiberboard overpack.
- Collapses as product is dispensed
- Non-glug design assures product flows from in continuous, uninterrupted stream
Customers Who Bought This Also Bought