ContactUsLinkComponent
Sulfenylated Protein Cell-Based Detection Kit, Cayman Chemical
Sulfenylated Protein Cell-Based Detection Kit, Cayman Chemical
# de catalogue 75816-980
Numéro CAS:  
Sulfenylated Protein Cell-Based Detection Kit
Sulfenylated Protein Cell-Based Detection Kit, Cayman Chemical
# de catalogue 75816-980
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75816-980
  • Contents:
    Kit contains: Daz-2, Buffer, Fixative, Blocking Solution, Phosphine-biotin, Complex, and EGCG
  • Supplier No.:
    600320-96
Spécifications
A propos de cet article
For the detection of sulfenic acid-modified proteins in living cells
  • Contains the cell-permeable probe DAz-2 to detect sulfenic acid-modified proteins in cells
  • Includes EGCG as a positive control
  • Detect by flow cytometry, fluorescence microscopy, or plate reader
Reactive oxygen species (ROS) react with proteins, resulting in protein modification, such as protein sulfenylation through the reversible oxidation of cysteine residues. Redox-sensitive cysteine residues in proteins can function as sensors of ROS and serve as molecular switches, activating or deactivating proteins, following a change in oxidative state. However, the accumulation of proteins with irreversible cysteine oxidation is a hallmark of stress-induced cellular damage associated with diseases like cancer. Cayman’s Sulfenylated Protein Cell-Based Detection Kit employs the cell-permeable and chemoselective DAz-2 probe to detect sulfenic acid-modified proteins in living cells. It can be used to monitor intracellular sulfenylated protein levels in living cells to discriminate between normal and pathological conditions or to help identify new pathways regulated by sulfenic acid formation.
Customers Who Bought This Also Bought