A propos de cet article
A fluorometric detection assay for hydrogen peroxide
- A simple fluorometric assay for the quantification of extracellular H2O2
- Utilizes ADHP, a sensitive and stable probe for H2O2
- Catalase, an H2O2 scavenger, is included as a control for specificity
- Rapid assay; results in under 1 hour
- Midpoint: 50% B/B0: 100-300 pg/ml
Reactive oxygen species, such as hydrogen peroxide (H2O2) and superoxide are generated by phagocytes and participate in damaging invading miccroorganisms or other biologic targets. Cayman's H2O2 Cell-Based Assay Kit provides a simple fluorometric method for the sensitive quantitation of extracellular H2O2 produced by cultured cells. H2O2 is detected using ADHP (10-acetyl-3,7-dihydroxyphenoxazine), a highly sensitive and stable probe for H2O2. In the presence of horseradish peroxidase, ADHP reacts with H2O2 with a 1:1 stoichiometry to produce highly fluorescent resorufin. Resorufin fluorescence can be read using an excitation wavelength between 530-560 nm and an emission wavelength of 590 nm. Catalase, an H2O2 scavenger, is included in the kit for checking specificity of the assay.