ContactUsLinkComponent
S-Glutathionylated Protein Detection Kit, Cayman Chemical Company
S-Glutathionylated Protein Detection Kit, Cayman Chemical Company
# de catalogue 75816-760
Numéro CAS:  
KIT DETCN S-GLUTATHIONYLATED PRT 30 SAMP
S-Glutathionylated Protein Detection Kit, Cayman Chemical Company
# de catalogue 75816-760
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75816-760
  • Contents:
    Kit contains: Buffers and Reagents
  • Supplier No.:
    10010721-1
Spécifications
A propos de cet article
A convenient method for the direct visualization of S-glutathionylated proteins in whole (permeabilized) cells
  • Direct visualization of S-glutathionylated proteins in whole (permeabilized) cells by flow cytometry and microscopy
  • Visualization using colorimetric or fluorescence detection
  • Reagents provided to test up to 30 samples
Glutathione (GSH) is involved in maintenance of protein sulfhydryl reduction status. PSSG may modify the activity of a large number of cell proteins, including metabolic, structural, cytoskeletal, and signaling proteins. Cayman’s PSSG provides a convenient method for the direct visualization of S-glutathionylated proteins in whole (permeabilized) cells by flow cytometry and microscopy as well as avidin overlay analysis. This cell-based assay starts with the modification of protein free-thiols groups followed by enzymatic cleavage of any PSSG adducts present in the sample. Biotinylation of the newly-formed protein free-thiols provides the basis for visualization using streptavidin-based colorimetric or fluorescence detection. Reagents are provided to test three sets of 10 samples (most convenient) or up to thirty samples total at once if desired.
Customers Who Bought This Also Bought