ContactUsLinkComponent
Glutathione Cell-Based Detection Kit (Blue Fluorescence), Cayman Chemical Company
Glutathione Cell-Based Detection Kit (Blue Fluorescence), Cayman Chemical Company
# de catalogue 75816-984
Numéro CAS:  
KIT DETCN 96 WELL GTN CEL BSD BLUE FLUOR
Glutathione Cell-Based Detection Kit (Blue Fluorescence), Cayman Chemical Company
# de catalogue 75816-984
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75816-984
  • Contents:
    Kit contains: Monochlorobimane, Standard, Buffer, and Plate
  • Supplier No.:
    600360-96
Spécifications
A propos de cet article
For rapid and easy determination of GSH in whole cells
  • Utilizes MCB, a highly fluorescent GSH probe
  • Includes a standard curve for accurate quantification of GSH from cell lysates
  • Rapid assay; get results in 2 hours
Glutathione (GSH) is a low molecular-weight tripeptide thiol that serves as a primary cellular anti-oxidant and plays a fundamental role in the elimination of environmental toxins. During early apoptosis, cells may exclude GSH, causing a decrease in intracellular GSH levels. The intracellular level of GSH is thus used as an indicator of cell health. Cayman’s Cell-Based Glutathione Assay Kit (Blue Fluorescence) employs a cell-permeable dye, monochlorobimane (MCB), which reacts with GSH to generate a highly fluorescent product that can be measured using excitation and emission wavelengths of 380 and 480 nm, respectively. The kit can be easily adapted to screening programs for therapeutic compounds regulating intracellular GSH levels.
Customers Who Bought This Also Bought