Order Entry
ContactUsLinkComponent
Ascorbate Assay Kit, Cayman Chemical
Ascorbate Assay Kit, Cayman Chemical
# de catalogue 75817-138
Numéro CAS:  
KIT ASCORBATE ASSAY 5-150 UM 96 WELLS
Ascorbate Assay Kit, Cayman Chemical
# de catalogue 75817-138
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Brand name:
    Cayman
  • Cat. no.:
    75817-138
  • Contents:
    Kit contains: Standard, Buffer, DTPA, Substrate, Developer, and Pate
  • Range:
    5 - 150 µM
  • Supplier No.:
    700420-96

Spécifications

A propos de cet article

A kit for measuring ascorbate

  • Quantify ascorbate from plasma, serum, urine, and fruit juices
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 340-350 nm, em 420-430 nm)

Ascorbate (L-Ascorbic acid or Vitamin C) is a six-carbon lactone that is synthesized from glucose in the liver of most mammalian species, but not by humans who must obtain ascorbate through dietary consumption. In humans, ascorbate serves as an antioxidant and acts as an electron donor for various enzymes. Cayman’s Ascorbate Assay provides a reproducible, sensitive fluorescence-based tool for quantifying ascorbate from plasma, serum, urine, and fruit juices.

Customers Who Bought This Also Bought