ContactUsLinkComponent
NOS Activity Assay Kit, Cayman Chemical Company
NOS Activity Assay Kit, Cayman Chemical Company
# de catalogue 75817-234
Numéro CAS:  
KIT NOS ACTIVITY ASSAY SENSITIVE RT
NOS Activity Assay Kit, Cayman Chemical Company
# de catalogue 75817-234
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75817-234
  • Contents:
    Kit contains: Positive Control, Buffer, Reagents, Spin Columns, and Collection Tubes
  • Supplier No.:
    781001-1
Spécifications
A propos de cet article
For the measurement of NOS activity
  • Measure NOS activity in cell lysates and purified preparations
  • Material for 50 reactions
  • Radioactive substrate not supplied
The NOS Activity Assay measures NOS activity by monitoring the conversion of radiolabeled arginine to citrulline. This assay is simple, sensitive, and specific for NOS activity and can be used with both crude and purified enzyme preparations. The kit includes sufficient materials and reagents for 50 total reactions. Radiolabeled arginine and NADPH are not included with the kit.
Customers Who Bought This Also Bought