ContactUsLinkComponent
Cholesterol Cell-Based Detection Assay Kit, Cayman Chemical Company
Cholesterol Cell-Based Detection Assay Kit, Cayman Chemical Company
# de catalogue 75816-742
Numéro CAS:  
KIT DETCN ASY 192 WL CHOLESTEROL CEL-BSD
Cholesterol Cell-Based Detection Assay Kit, Cayman Chemical Company
# de catalogue 75816-742
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75816-742
  • Contents:
    Kit contains: Buffer, Fixative, Fillipin III, and U-18666A
  • Supplier No.:
    10009779-192
Spécifications
A propos de cet article
A 192-well assay for measurement of cholesterol
  • Detect cholesterol uptake and localization within cultured cells
  • Employs filipin III, a probe that fluoresces in the presence of cholesterol
  • An inhibitor of cholesterol trafficking is included as a positive control
The mechanism for the movement of cholesterol from intracellular sites to their ultimate cellular destination is an unresolved question of fundamental importance to cell biology and medicine. Thus, defining mechanisms of intracellular cholesterol transport and identifying the cellular factors involved are therefore of great interest. Cayman’s Cholesterol Cell-Based Detection Assay includes filipin III, fixative, and wash buffer in a ready to use format. It provides a simple fluorometric method to study mechanisms and biological factors that regulate cholesterol metabolism or movement within cells. A cholesterol trafficking inhibitor, U-18666A, is included as a positive control.
Customers Who Bought This Also Bought