ContactUsLinkComponent
Oxygen Consumption Rate Assay Kit, Cayman Chemical
Oxygen Consumption Rate Assay Kit, Cayman Chemical
# de catalogue 75817-070
Numéro CAS:  
Oxygen Consumption Rate Assay Kit (MitoXpress* Xtra HS Method)
Oxygen Consumption Rate Assay Kit, Cayman Chemical
# de catalogue 75817-070
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Brand name:
    Cayman
  • Cat. no.:
    75817-070
  • Contents:
    Kit contains: Probe and Reagents
  • Supplier No.:
    600800-96
Spécifications
A propos de cet article
For the measurement of oxygen consumption rate in living cells
  • Measure OCR without the need for an oxygen electrode
  • Includes antimycin A, an inhibitor of oxygen consumption, as a control
  • Includes glucose oxidase as a reference for oxygen depletion
The oxygen consumption rate (OCR) of cells is an important indicator of normal cellular function. It is used as a parameter to study mitochondrial function as well as a marker of factors triggering the switch from healthy oxidative phosphorylation to aerobic glycolysis in cancer cells. Oxygen consumption is traditionally measured by a cumbersome oxygen electrode, a specialized piece of equipment that typically yields low sample throughput. A Phosphorescent Oxygen Probe has proven to be useful in measuring oxygen consumption rates in whole cells. Cayman’s cell-based Oxygen Consumption Rate Assay Kit utilizes this newly developed phosphorescent oxygen probe to measure oxygen consumption rate in living cells. Antimycin A, an inhibitor of the mitochondrial electron transport chain, is included to be used as a positive control. Glucose oxidase is also included in the kit to be used as a reference for oxygen depletion. The kit is easy to use and can be easily adapted to high throughput screening for compounds which modulate oxygen consumption rate.
Customers Who Bought This Also Bought