A propos de cet article
For the quantification of oxytocin in plasma
- Measure oxytocin in plasma
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure oxytocin levels down to 20 pg/ml
- Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
- Midpoint: 50% B/B0: 50-100 pg/ml
Oxytocin is a hypothalamic peptide hormone that is stored in the posterior pituitary gland. It is released into the bloodstream during parturition and lactation and is also involved in other social and sexual behaviors, neuropsyciatric disorders, and the maintenance of water and sodium homeostasis. Cayman’s Oxytocin ELISA Kit is a competitive assay that can be used for quantification of oxytocin in plasma. The assay has a range from 5.9-750 pg/ml and a sensitivity (80% B/B0) of approximately 20 pg/ml.