Order Entry
ContactUsLinkComponent
L-Lactate Assay Kit, Cayman Chemical
L-Lactate Assay Kit, Cayman Chemical
# de catalogue 75817-146
Numéro CAS:  
KIT L-LACTATE ASSAY 25 UM-1 MM 96 WELLS
L-Lactate Assay Kit, Cayman Chemical
# de catalogue 75817-146
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Brand name:
    Cayman
  • Cat. no.:
    75817-146
  • Contents:
    Kit contains: Buffer, Mixtures, Substrate, Standard, Reagents, and Plate
  • Range:
    25 μM - 1 mM
  • Supplier No.:
    700510-96

Spécifications

A propos de cet article

A fluorescence-based method for detecting L-lactate in biological samples such as serum, plasma, blood, urine, and saliva

  • Measure L-lactate in biological samples such as serum, plasma, whole blood, urine, saliva, and cell culture
  • Assay 40 samples in duplicate
  • Plate-based fluorometric measurement (ex 530 – 540 nm, em 585 – 595 nm)

L(+)-Lactate is the major stereoisomer of lactate formed in human intermediary metabolism. The lactate to pyruvate ratio reflects the redox state of the cell and describes the balance between NAD+ and NADH, which is dependent on the interconversion of lactate and pyruvate via lactate dehydrogenase (LDH). Monitoring lactate levels is, therefore, a good way to evaluate the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Cayman’s L-Lactate Assay provides a fluorescence-based method for detecting L-lactate in biological samples such as serum, plasma, blood, urine, and saliva. It can also be utilized to determine intracellular and extracellular lactate concentrations in cell culture samples. In the assay, lactate dehydrogenase catalyzes the oxidation of lactate to pyruvate, along with the concomitant reduction of NAD+ to NADH. NADH reacts with the fluorescent substrate to yield a highly fluorescent product. The fluorescent product is analyzed with an excitation wavelength of 530-540 nm and an emission wavelength of 585-595 nm.

Customers Who Bought This Also Bought